Learn More
Andwin Scientific Direct Thermal Cryo-Tags, 1.50X 0.7inch, Blue, 750/roll
SDP
Supplier:  Andwin Scientific DTCR3000B
Encompass_Preferred
Direct Thermal Cryo-Tags are formatted to easily work with the Dymo Labelwriter, but also with most direct thermal printers. Quickly label your samples with sharp text, clear barcodes or a combination of both. Temperatures From 70 to -196degreeC.
Catalog No. NC1659343
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.