Learn More
DTNA Mouse anti-Human, Clone: OTI1C2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF502164

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene belongs to the dystrobrevin subfamily of the dystrophin family. This protein is a component of the dystrophin-associated protein complex (DPC), which consists of dystrophin and several integral and peripheral membrane proteins, including dystroglycans, sarcoglycans, syntrophins and alpha- and beta-dystrobrevin. The DPC localizes to the sarcolemma and its disruption is associated with various forms of muscular dystrophy. Mutations in this gene are associated with left ventricular noncompaction with congenital heart defects. Multiple alternatively spliced transcript variants encoding different isoforms have been identified for this gene.Specifications
DTNA | |
Monoclonal | |
Unconjugated | |
DTNA | |
D18S892E, DRP3, DTN, DTN-A, LVNC1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
1837 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI1C2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9Y4J8 | |
DTNA | |
Full length human recombit protein of human DTNA produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.