Learn More
DTNB Mouse anti-Human, Mouse, Rat, Clone: OTI3H8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF504332

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes dystrobrevin beta, a component of the dystrophin-associated protein complex (DPC). The DPC consists of dystrophin and several integral and peripheral membrane proteins, including dystroglycans, sarcoglycans, syntrophins and dystrobrevin alpha and beta. The DPC localizes to the sarcolemma and its disruption is associated with various forms of muscular dystrophy. Dystrobrevin beta is thought to interact with syntrophin and the DP71 short form of dystrophin. Alternatively spliced transcript variants encoding different isoforms have been identified.Specifications
DTNB | |
Monoclonal | |
Unconjugated | |
DTNB | |
dystrobrevin beta | |
Mouse | |
Affinity Chromatography | |
RUO | |
13528, 1838, 362715 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI3H8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O60941, O70585, P84060 | |
DTNB | |
Full length human recombit protein of human DTNB produced in HEK293T cell. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.