Learn More
Abcam DTNB, Sulfhydryl Reagent, 99 Percent Purity, Solid, 5 g
Description
- Molecular weight: 396.4 g/mol
- Purity: >99%
- Form: Solid
- Composition: 100% 5,5'-Dithiobis(2-nitrobenzoic acid)
- Application: Sulfhydryl reagent for characterizing reactive thiol groups, including photometric determination of thiols and HPLC analysis of alkylthiol content
- Storage: Store at +4°C
- Shipping: Ambient - Can Ship with Ice
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Sulfhydryl Groups |
| Substrate Properties | Chemical Substrate |
| Substrate | Sulfhydryl groups |
| Shipping Condition | Ambient |
| Buffer | 100% 5,5'-dithiobis(2-nitrobenzoic acid) |
| Content And Storage | Store at 4°C. |
| Form | Solid |
| Detection Method | Colorimetric |
| Formulation | 100% 5,5'-Dithiobis(2-nitrobenzoic acid) |
| Quantity | 5 g |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.