Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DTWD2 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. p-7210184 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4588 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4588 Supplier Proteintech Group Inc Supplier No. 248731AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DTWD2 is a protein coding gene.

Specifications

Antigen DTWD2
Applications Immunoprecipitation
Classification Polyclonal
Concentration 0.32 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene DTWD2
Gene Accession No. Q8NBA8
Gene Alias DTW domain containing 2, DTWD2
Gene Symbols DTWD2
Host Species Rabbit
Immunogen DTWD2 Fusion Protein Ag21665
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285605
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.