Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBF Bioscience Dual Cool Mini-Vertical PAGE/Blotting System
SCI_ED

Catalog No. S05759 Shop All CBS Scientific Products
Change view
Click to view available options
Device Holds:
0.75 mm x 10-well Precast Gel
1.5 mm x 10-well Precast Gel
1.5 mm x 14-well Precast Gel
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Device Holds
S05759 1.5 mm x 14-well Precast Gel
S05758 1.5 mm x 10-well Precast Gel
S05754 0.75 mm x 10-well Precast Gel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S05759 Supplier MBF Bioscience Supplier No. DCXHC1514

missing translation for 'validMainframeMsg'

DCX-700 with Gel Wrap™ Handcasting Bundle DCX-HC.

  • Uniquely designed, multipurpose, easy-to-use Dual Cool System can accommodate both slab gel electrophoresis and Western blotting within the same unit without the purchase of additional modules
  • The Dual Cool System provides the capability of running 2 gels or 2 blots simultaneously under identical temperature controlled buffer conditions
  • DCX-700 Dual Cool Electrophoresis System includes lower reservoir, core, 2 blotting cassettes, 2 freezer blocks, safety cover with attached leads, single gel adapter plate
  • Handcasting components include two sets of Gel Wrap glass plates, 2 sets of Gel Wrap spacers (choice of 0.75mm, 1.0mm, or 1.5mm thicknesses), 2 combs (choice of 0.75mm, 1.0mm or 1.5mm thickness with 10 or 14 wells), 2 Gel Wrap gaskets for sealing glass plate sandwich (choice of 0.75mm, 1.0mm and 1.5mm) and 6 casting clamps

DCX-700

  • Molded system is designed primarily for use with a variety of precast gels ranging in size from 10cm x 10cm to 10cm x 9cm
  • Alternatively, hand-cast gels can be run with the use of accessory combs, plates, and spacers
  • Hinged core design allows easy loading and removal of gel or blotting cassettes
  • Freezer blocks fit into receptacles inside of unit for optional cooling
  • Electrophoretic transfers do not require the purchase of separate transfer modules
  • Color-coded anode and cathode safety features on all components
  • Clear tank allows for monitoring of sample progress
  • Platinum electrodes

SDS-PAGE, Electroblotting (Western), Acrylamide nucleic acid separations

Certifications

CE marked

Specifications

Ambient Temperature (English) 140°F
Ambient Temperature (Metric) 60°C
Certifications/Compliance EN61010-1-1993 (IEC1010-1), CE
Device Holds 1.5 mm x 14-well Precast Gel
Type Dual Cool Mini Vertical PAGE/Blotting System
Includes Gel Wrap™ handcasting bundle, Two 1.5mm x 14-well combs, 2 sets 1.5mm spacers, 2 sets 1.5mm Gel Wrap™ gaskets, 2 sets Glass plates and 6 clamps
Volume (English) Buffer 13.52 to 27.38 oz.
Volume (Metric) Buffer 400 to 810 mL
Dimensions (L x W x H) 5.98 x 6.49 x 8.5 in. (15.2 x 16.5 x 21.6 cm)
Length (English) 5.98 in.
Length (English) Gel 3.93, 3.54, 3.14 in.
Length (Metric) 15.2 cm
Length (Metric) Gel 10, 9, 8 cm
Width (English) 6.49 in.
Width (English) Gel 3.93 in.
Width (Metric) 16.5 cm
Width (Metric) Gel 10 cm
Height (English) 8.5 in.
Height (Metric) 21.6 cm
Voltage 300 V
Wattage 30 w
Amperage 190 mA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.