Learn More
Fibre-Metal by Honeywell™ Dual-Crown High-Performance Faceshield
Catalog No. 19070302 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
4 in.3C crown with clear chin protector
Catalog No.
19-070-302 4 in.3C crown with clear chin protector
1 options
This item is not returnable. View return policy
Catalog No. 19-070-302 Supplier Fibre-Metal by Honeywell™ Supplier No. FM400DCCLC
Add to Cart
Add to Cart
This item is not returnable. View return policy
Dual crown headgear and faceshield provides wraparound protection.
  • Dual crown high performance faceshield
  • 4 in. deep chin protectors in clear polycarbonate material
  • 3-C ratchet
  • Accepts all Quick-Lok™ and Speedy™ Loop accessories
  • SEI certified
Certifications
SEI certified
Compliance
Meets ANSI-Z87.1+ 2003 standards.
Specifications
Includes F400 4 in. crown with 6750CL clear window & clear chin guard complete, 3C ratchet