Learn More
United Scientific Supplies Dual Drive Van de Graaff Generator
SDPSCI_ED
Catalog No. S27874 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Product Type:
Replacement Belt
Van De Graaff Generator, Dual Drive
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S27874 Replacement Belt
S27873 Van De Graaff Generator, Dual Drive
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S27874 Supplier United Scientific Supplies Supplier No. VANDGF1RB
Add to Cart
Add to Cart
Uniquely offers hand-cranked or motor driven alternative drive modes
  • Operates by hand crank or by motor
  • Generates approximately 200,000 volts
  • Produces immediate and lengthy sparks, even in humid conditions
  • Collector sphere crafted from dent- and scratch-resistant polished steel
  • Built-in UV lamp provides additional light
  • Includes accessory kit and activity guide
  • Replacement Belt sold separately
  • Diameter: 28 cm (Collector Sphere)
  • Device: 38 x 23 x 76 cm
Specifications
Product Type Replacement Belt
For Use With (Equipment) Van De Graaff Generator, Dual Drive
Quantity 1