Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allegro™ Dual-Flex Wrist Support

Catalog No. 19037866B
Change view
Click to view available options
Size:
Small
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-037-866B Small
19-037-866D Large
19-037-866E X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-037-866B Supplier Allegro™ Supplier No. 721201
Add to Cart
Add to Cart

Specifications

Color Black
Closure Type Hook and loop
Product Type Dual-Flex™ Wrist Support
Size Small
For Use With (Application) Offers firm support limit flexion (downward) and extension (upward) movement and encourages natural wrist position
Includes Four stays on the front and back
Hand Ambidextrous
Material Nylon
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.