Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dual Hand Lens
SCI_ED

Catalog No. S93920 Shop All Si Metric Manufacturing Limited Products
Change view
Click to view available options
Magnification:
3, 5x
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Magnification
S93920 3, 5x
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S93920 Supplier Si Metric Manufacturing Limited Supplier No. 14434

missing translation for 'validMainframeMsg'

Versatile.

  • 3x and 5x magnifers
  • 0.75 and 1.5 in. diameter lens

Order Info

Grades K+.

Specifications

Diameter (Metric) Lens 1.91, 3.81 cm
Product Type Lens, Dual Hand
Diameter (Englis) Lens 0.75, 1.5 in.
Magnification 3, 5x
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.