Learn More
Bioassay Systems SuperLight™ Dual Luciferase Reporter Gene Assay Kit. Bioluminescent reagent system for rapid quantitation of firelfy and Ranilla luciferase reporter gene expression in transfected cells.
SDP
Catalog No. 50489302 Shop All Bioassay Systems Products
Encompass_Preferred
Bioluminescent reagent system for rapid quantitation of firelfy and Ranilla luciferase reporter gene expression in transfected cells. Key Features: High sensitivity and wide detection range. Detection of as little of 2 fg luciferase. Compatible with routine laboratory and HTS formats. Assays can be performed in tubes or microplates, and measured with any luminometer. Can be readily automated on HTS liquid handling systems. Fast and convenient. Three step assay allows detection of dual luciferase levels within 20 minutes. Method: Luminescence. Samples: Cells etc. Species: All. Procedure: Assay takes 20 min. Kit size: 100 tests. Detection limit: 2 fg luciferase.
Catalog No. 50-489-302 Supplier Bioassay Systems Supplier No. SLDL100
Bioluminescent reagent system for rapid quantitation of firelfy and Ranilla luciferase reporter gene expression in transfected cells. Key Features: High sensitivity and wide detection range. Detection of as little of 2 fg luciferase. Compatible with routine laboratory and HTS formats. Assays can be performed in tubes or microplates, and measured with any luminometer. Can be readily automated on HTS liquid handling systems. Fast and convenient. Three step assay allows detection of dual luciferase levels within 20 minutes. Method: Luminescence. Samples: Cells etc. Species: All. Procedure: Assay takes 20 min. Kit size: 100 tests. Detection limit: 2 fg luciferase.