Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PureAire Monitoring Systems Dual O2/CO2 Monitor 0 to 25 and 0 to 50,000 ppm
SDP

Catalog No. 17377543
Change view
Click to view available options
Detectable Analytes:
Oxygen
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes
17-377-543 Oxygen
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-377-543 Supplier PureAire Monitoring Systems Supplier No. 99143
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PureAire Monitoring System's Dual O2/CO2 Monitor provides continuous monitoring of oxygen and carbon dioxide. This is ideal for use in confined spaces where low oxygen concentrations could pose a health hazard, from nitrogen gas storage sites to carbon dioxide beverage gas distribution areas.

  • No scheduled maintenance zirconium O2 cell and long-life CO2 sensor
  • No calibration is required for oxygen
  • Self-calibrating CO2 sensor
  • Local backlit concentration display
  • Built-in audible horn
  • Built-in visual LED alarm indicators
  • Alarm level 1 for CO2 / Alarm level 2 for O2
  • No drift due to environmental or temperature changes
  • 3-year warranty
  • 24n V DC Power Supply Included

Specifications

Detectable Analytes Oxygen
Detection Limits Oxygen 0 to 25%/CO2 0 to 50,000 ppm
For Use With (Equipment) Optional equipment-PureAire Horn/Strobe, Remote Digital Display, Multi-Channel Controller
Inputs 0
Product Type Air Monitoring System
Response Time 15 sec. (1 sec. average time)
Temperature (Metric) Operating -40°C to +60°C
Accuracy Delivers ±0.2% O2 accuracy (± 1% of full range) /CO2 ±300 ppm, ± 3% of measured value
Alarm Averaging Time Within 1 second of any change in O2 or CO2
Alarms Audible 90 dB/Visual-Orange LED alarm indicator for CO2, Red LED alarm indicator for O2
Amperage 250 mA
Certifications/Compliance CE
Concentration Measurement Range O2 0 to 25%, CO2 0 to 50,000 ppm
Depth (English) 3.25 in.
Depth (Metric) 82.5 mm
Detection Range Oxygen 0 to 25%, Carbon Dioxide 0 to 50,000 ppm
Dimensions (L x W x H) 6.5 in. x 4.5 in. x 3.25 in.
Display Type 0.75 in. backlit LCD digital display
Electrical Requirements 24 VDC external power
Enclosure Material Polycarbonate-General Purpose
For Use With (Application) To detect and warn of unsafe oxygen and/or carbon dioxide levels
Height (English) 4.5 in.
Height (Metric) 114.3 mm
Includes 24 V Power Supply
Length (English) 6.5 in.
Length (Metric) 165.1 mm
Measurement Range Oxygen 0 to 25%/Carbon Dioxide 0 to 50,000 ppm
Mounting Options Wall Mount
Outputs One user-adjustable Alarm relay for Oxygen and one user-adjustable Alarm relay for CO2. Relays are rated 2 amp 24 VDC / 24 VAC
Precision Delivers ± 0.2% O2 accuracy (± 1 % of Full Range)-O2
Pressure ±300 ppm, ± 3% of measured value - CO2
Real-time Analog Signal Analog 4-20 mA output (Active)
Sensing Element Type Oxygen -Zirconium Oxide Sensor Cell/Carbon Dioxide -NDIR sensor cell
Voltage 24 V
Weight (English) 1.6 lb.
Weight (Metric) 0.8 kg
Width (English) 3.25 in.
Width (Metric) 82.5 mm
Measurement Method Diffusion
Temperature (English) Operating -40°F to +140°F
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.