Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labcon Dual Position Caps for 12 x 75 mm Culture Tubes, in Bags

Catalog No. 50101612
Encompass
Encompass_Preferred
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
50-101-612 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-101-612 Supplier Labcon Supplier No. 33008000009
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dual position aspirating cap for culture tubes. Fit popular 12 x 75 mm culture tubes.

  • Cap includes plug position and open aspirating position
  • Endotoxin Free (non-pyrogenic)
  • Polyethylene USP Class VI resin
  • Natural white color
  • Made in USA

Specifications

Certifications/Compliance ISO 9001:2015; FDA CFR21 GMP
Product Type Dual-position Cap
Closure Size 13 mm
Pre-sterilized No
Autoclavable Not Autoclavable
Material Polyethylene
Color Clear
For Use With (Application) Centrifugation; For Cell Culture and General Laboratory Use
No. per Pack 1000
No. per Case 3000
Clean Claims ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
Sterility Non-sterile
For Use With (Equipment) Tubes
Disposable Single-use
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.