Learn More
Electron Microscopy Sciences Rotatrim Rolling Trimmer Dual-Rail Cutter, 36"
SDP
Supplier:  Electron Microscopy Sciences 7488036
Encompass_Preferred
For precision cutting through tough materials like cardboard, litho plates, mount board, and even 1/16" hardboard. Rotatrim also has a delicate touch, trimming film, paper, and dry mount tissue to hairline tolerances.
Known for its safe operation and best performance, the rotary blade is Sheffield tungsten steel, self-sharpening, and is completely shielded by a protective housing. Its self-sharpening blade assures consistent cutting quality.
Exclusively from Rotatrim, its double steel tube suspension guides the cutting head with the stability essential for accurate, repeatable cuts. An extruded aluminum squaring arm incorporates a combination inch/metric measuring scale and accepts the accessory side guide. A Transparent clamping strip, holds material in place without risk of damage.
Its durable, high pressure laminate finish keeps the baseboard warp-free, long lasting, and easy to clean.
Size: 36"
Catalog No. 50-283-93
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.