Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier™ Dual-Range Force Sensor
SDPSCI_ED

Catalog No. S16050ND Shop All Vernier Software Products
Change view
Click to view available options
Grade:
K-8, High School, College
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S16050ND K-8, High School, College
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S16050ND Supplier Vernier™ Supplier No. DFSBTA
Add to Cart
Add to Cart

Includes

Dual-Range Force Sensor, Hook, Bumper, Nylon thumb screw, Accessory Rod

Requires

Requires an interface and software, sold separately

Use the Dual-Range Force Sensor in a wide variety of experiments, including the study of friction, simple harmonic motion, impact in collisions, or centripetal force.

± 10 N Range Resolution: 0.01 N ± 50 N Range Resolution: 0.05 N

Warranty and Services

5-year limited warranty

Specifications

Grade K-8, High School, College
Interface Required
Warranty 5 Year Limited
For Use With (Application) Educational use only
Resolution ± 10 N Range Resolution: 0.01 N; ± 50 N Range Resolution: 0.05 N
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.