Learn More
Kingfisher Biotech Inc Mallard IL-4 Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP1743MD005
Encompass
The Duck IL-4 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Duck IL-4 applications are for cell culture, ELISA standard, and Western Blot Control. Duck IL-4 yeast-derived recombinant protein can be purchased in multiple sizes. Duck IL-4 Specifications: (Molecular Weight: 12.8 kDa) (Amino Acid Sequence: HEAAVLERSI FLKESIRLLG EILSTQVSCD KANVTNVFAG NETGTDMELL CKASTVVFES LSCHKPLKGI YLNLLHIVTK STSLKAPCPV AAGNTTSLQE FLRGLHRTLQ RVAKENL (117)) (Gene ID: 101801081). For research use only. Made in the USA
Catalog No. 50-253-2845
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.