Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ ProShield™ 60 Coverall (all)

Catalog No. 190882153 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Height (English):
5 to 5.7 ft.
5.3 to 5.7 ft.
5.5 to 5.9 ft.
5.8 to 6.2 ft.
6 to 6.4 ft.
6.2 to 6.4 ft.
6.4 to 6.7 ft.
6.7 to 6.10 ft.
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Height (English)
19-088-2153 Large 5.5 to 5.9 ft.
19-065-172 5X-Large 6.7 to 6.10 ft.
19-088-2151 Small 5 to 5.7 ft.
19-088-2154 X-Large 5.8 to 6.2 ft.
19-088-2156 3X-Large 6.2 to 6.4 ft.
19-088-2152 Medium 5.3 to 5.7 ft.
19-088-2157 4X-Large 6.4 to 6.7 ft.
19-088-2155 2X-Large 6 to 6.4 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 19-088-2153 Supplier DuPont™ Supplier No. NG125SWHLG002500
This item is not returnable. View return policy

Coveralls are suitable for protection against non-hazardous particles, light liquid splash, and aerosols in many industrial applications.

  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Laydown collar
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Manufactured under specifications that do not contain natural rubber latex

  1. Measured per ASTM™ F 903 in relation to the following substances: household bleach, diazinon (4 oz./gal.), hydraulic jack oil, machine oil, malathion (50%), motor oil (40W), sodium hydroxide and isocyanate-based paint hardener. Tests were performed by independent laboratories on material samples under laboratory conditions, not with actual garments or in actual onsite conditions.
  2. ASTM D 751 (Grab Tensile Strength), ASTM D 1117 (Trapezoid Tear), ASTM D 3787 (Ball Burst).
  3. Certified incinerable by independent laboratory. For details, contact your Fisher Safety Customer Service Representative at 1-800-772-6733.

TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Coveralls
Size Large
Gender Unisex
Material ProShield
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Hood Type Unhooded
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Particle, Liquid
Collar Type Laydown
Certifications/Compliance ASTM D751, D1117, D5034, D257, AATCC 127
Description Microporous Film, Laminated to a Spundbonded Polypropylene Substrate
Latex Free Yes
Model NG125S
Size (English) Chest 37.25 to 40.75 in.
Size (English) Inseam 27 in.
Size (Metric) Chest 94.61 to 103.5 cm
Size (Metric) Inseam 68.58 cm
Thickness 10 mil
For Use With (Application) Automotive Refinishing, Waste Cleanup and Remediation
Length (English) Sleeve 32.75 in.
Length (Metric) Sleeve 83.18 cm
Height (English) 5.5 to 5.9 ft.
Height (Metric) 1.67 to 1.8 m
Quantity 25/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.