Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ ProShield™ 10 Hooded Coverall

Catalog No. 19162651 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
7X-Large
Color:
Blue
White
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Color
19-162-651 7X-Large White
19-162-549 Medium Blue
19-162-550 Large Blue
19-162-551 X-Large Blue
19-162-552 2X-Large Blue
19-162-553 3X-Large Blue
19-162-554 4X-Large Blue
19-162-555 5X-Large Blue
19-162-645 6X-Large Blue
19-162-652 7X-Large Blue
19-162-542 Medium White
19-162-543 Large White
19-162-544 X-Large White
19-162-546 3X-Large White
19-162-547 4X-Large White
19-162-548 5X-Large White
19-162-646 6X-Large White
19-162-545 2X-Large White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
This item is not returnable. View return policy
Catalog No. 19-162-651 Supplier DuPont™ Supplier No. PB127SWH7X002500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Spunbond meltblown spunbond (SMS) coveralls protect against non-hazardous particles and light liquid splashes in a variety of industries.

  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Attached hood with elastic around face opening.
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Manufactured under specifications that do not contain natural rubber latex
TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Coveralls
Size 7X-Large
Gender Unisex
Material ProShield
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Particle
Latex Free Yes
Model PB127SWH
For Use With (Application) General Maintenance, Janitorial, Cleaning
Quantity 25/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.