Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ ProShield™ 10 Frock

Catalog No. 19162636 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-162-636 Large
19-162-637 X-Large
19-162-639 3X-Large
19-162-638 2X-Large
19-162-640 4X-Large
19-162-635 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-162-636 Supplier DuPont™ Supplier No. PB267SWHLG003000
Add to Cart
Add to Cart
This item is not returnable. View return policy

SMS frocks are designed to be used in a range of industries for nonhazardous dry particle and light splash applications.

  • Frock provides body-length jacket design with front snap closures (6+1 adjustable) for easy donning and doffing
  • Mandarin collar
  • Extends to knee
  • Set sleeve
  • Knit cuff
  • Enclosed elastic wrist
  • Serged seams
  • Manufactured under specifications that do not contain natural rubber latex
TRUSTED_SUSTAINABILITY

Specifications

Size Large
Material ProShield
Color White
Disposable Single-use
Closure Type Snaps
Cuff Style Elastic
Collar Type Mandarin
Latex Free Yes
Product Type General Purpose Frock
Seam Type Serged
For Use With (Application) General Maintenance, Janitorial, Cleaning
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.