Learn More
DuPont™ ProShield™ 10 Lab Coat
Catalog No. 19162626 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Color:
Blue
White
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-162-626 White Small
19-162-619 Blue Small
19-162-620 Blue Medium
19-162-621 Blue Large
19-162-622 Blue X-Large
19-162-623 Blue 2X-Large
19-162-624 Blue 3X-Large
19-162-625 Blue 4X-Large
19-162-627 White Medium
19-162-628 White Large
19-162-629 White X-Large
19-162-630 White 2X-Large
19-162-631 White 3X-Large
19-162-632 White 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
This item is not returnable. View return policy
Catalog No. 19-162-626 Supplier DuPont™ Supplier No. PB219SWHSM003000
Add to Cart
Add to Cart
This item is not returnable. View return policy
SMS disposable lab coats are designed for a range of industries for nonhazardous dry particle and light liquid splash applications. Available in blue or white in a variety of sizes.
  • Knit collar
  • Set sleeve
  • Knit cuff
  • Manufactured under specifications that do not contain natural rubber latex
TRUSTED_SUSTAINABILITY
Specifications
Product Type General Purpose Lab Coat
Size Small
Gender Unisex
Material ProShield
Color White
Closure Type Snaps
Cuff Style Knit
Disposable Single-use
Sterility Non-sterile
No. of Pockets 3
Collar Type Knit
Latex Free Yes
Model PB219
Seam Type Serged
Certifications/Compliance ASTM D1777