Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tychem™ BR Coverall

Catalog No. 19067001 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
6X-Large
Height (English):
6 ft. 9 in. to 7 ft. 1 in.
Width (English) Chest:
35.75 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Height (English) Width (English) Chest
19-067-001 6X-Large 6 ft. 9 in. to 7 ft. 1 in. 35.75 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-067-001 Supplier DuPont™ Supplier No. BR127TYL6X000200
This item is not returnable. View return policy

Coverall with respirator fit hood, elastic wrists and ankles

Tychem™ BR offers wide-ranging protection backed by extensive fabric permeation barrier testing. Fabric provides at least 30 minutes of protection against more than 280 chemical challenges, including chemical warfare agents. It offers tear, puncture and abrasion resistant fabrics for lasting protection.
TRUSTED_SUSTAINABILITY

Specifications

Size 6X-Large
Material Tychem
Color Yellow
Seam Type Taped
Width (English) Chest 35.75 in.
Height (English) 6 ft. 9 in. to 7 ft. 1 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.