Learn More
DuPont™ Tychem™ 4000 Coverall (all)
Catalog No. 19813436 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Medium
X-Large
3X-Large
Height (English):
5.3 to 5.7 ft.
6.2 to 6.4 ft.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Height (English)
19-813-436 X-Large -
19-813-438 3X-Large 6.2 to 6.4 ft.
19-813-434 Medium 5.3 to 5.7 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-813-436 Supplier DuPont™ Supplier No. SL121TWHXL000400
Add to Cart
Add to Cart
This item is not returnable. View return policy
Tychem™ 4000 Coverall includes elastic wrists and attached socks.
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Laydown collar
  • Elastic opening for tighter fit at wrist
  • Integrated socks composed of garment material
  • Manufactured under specifications that do not contain natural rubber latex
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
TRUSTED_SUSTAINABILITY
Specifications
Size X-Large
Gender Unisex
Material Tychem
Color White
Ankle Style Attached Socks
Closure Type Zipper
Seam Type Taped