Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tychem™ 4000 Hooded Coverall

Catalog No. 19162775 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-162-775 Medium
19-162-769 3X-Large
19-162-768 2X-Large
19-162-770 4X-Large
19-162-772 5X-Large
19-162-774 Large
19-162-776 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Garments made of Tyvek™ should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Catalog No. 19-162-775 Supplier DuPont™ Supplier No. SL122TWHMD0006BN
This item is not returnable. View return policy
Garments made of Tyvek™ should not be used around heat, flames, sparks or in potentially flammable or explosive environments.

Delivers effective protection against a range of chemical environments.

  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Attached hood with elastic around face opening.
  • Elastic opening for tighter fit at wrist
  • Integrated socks composed of garment material
  • Manufactured under specifications that do not contain natural rubber latex
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Hooded Coveralls with Socks
Size Medium
Gender Unisex
Material Tychem
Color White
Disposable Single-use
Ankle Style Attached Socks
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached, Elastic Face
Closure Type Front Zipper
Seam Type Taped
Sterility Non-sterile
Description Attached hood, elastic wrist, attached socks
Size (English) Chest 35.25 to 38 .75 in.
Size (English) Inseam 27.5 in.
Size (Metric) Inseam 69.85 cm
Thickness 12 mil
For Use With (Application) Chemical mixing, hazardous remediation, emergency medical response, paint spraying and nuclear environments
Length (English) Sleeve 33.75 in.
Length (Metric) Sleeve 85.72 cm
Width (English) Chest 24.25 in.
Width (Metric) Chest 61.59 cm
Height (English) 5.25 to 5.58 ft.
Height (Metric) 1.58 to 1.7 m
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.