Learn More
DuPont™ Tychem™ 6000 FR Bib Overall And Jacket Combo
Catalog No. 19120962 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Color:
Gray
Orange
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Color
19-120-962 3X-Large Gray
19-120-957 Small Gray
19-120-959 Large Gray
19-120-960 X-Large Gray
19-120-961 2X-Large Gray
19-120-958 Medium Gray
19-120-963 4X-Large Gray
19-120-964 Small Orange
19-120-965 Medium Orange
19-120-966 Large Orange
19-120-967 X-Large Orange
19-120-970 4X-Large Orange
19-120-969 3X-Large Orange
19-120-968 2X-Large Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
This item is not returnable. View return policy
Catalog No. 19-120-962 Supplier DuPont™ Supplier No. TP750TGY3X000200
This item is not returnable. View return policy
Includes
  • Jacket with taped seams, mandarin collar, elastic wrists, front zipper closure and double storm flap
  • Bib Overall with taped seams and adjustable webbing straps with closure
Protection from chemicals, flash fire and electrical arc
  • Tychem™ Thermopro for triple hazard protection
  • Jacket and bib overall combination ensemble. Jacket and bib overall not sold separately.
  • Sewn with Nomex™ thread and over-taped with chemical barrier tape of same flame resistant composition as garment fabric
  • Key garment components (fabric, zipper, drawstrings) are flame resistant and thermally stable (will remain functional even after 5 min. exposure to 260°C)
  • Garment developed to meet the needs of industrial plant sites enabling easy donning and doffing
  • Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
  • Manufactured under specifications that do not contain natural rubber latex
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
TRUSTED_SUSTAINABILITY
Specifications
Color Gray
Material Tychem
Size 3X-Large
Ankle Style Open
Closure Type Zipper
Collar Type Mandarin
Cuff Style Elastic
Seam Type Taped