Learn More
Plasma Etch Inc DURABLE WELDED 6061 T6 CYLINDR
SDP
Supplier:  Plasma Etch Inc PE25JW
Encompass
Durable Welded 6061 T6 Cylindrical Aluminum Vacuum Chamber Dimensions: 5” ID x 8” Deep.
Catalog No. NC2865634
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.