Learn More
DWK Life Sciences DURAN™ Allihn Condenser with NS 29/32 Jacket
SDP
Catalog No. 09841074 Shop All DWK Life Sciences Products
Change view
Click to view available options
Length (Metric) Jacket:
250 mm
Catalog No. Length (Metric) Jacket
09-841-074 250 mm
1 options
Catalog No. 09-841-074 Supplier DWK Life Sciences Supplier No. 242527207
Add to Cart
Add to Cart
The DURAN™ Allihn Condenser with NS 29/32 Jacket features 2 standard ground joints and 2 screw-on plastic hose connections (GL 14 thread). Suited for distillate separations, this borosilicate glass condenser offers reliable cooling performance in laboratory applications.
  • DURAN™ borosilicate glass construction for chemical resistance and durability
  • NS 29/32 socket and cone standard ground joints
  • 2 screw-on plastic hose connections with GL 14 thread
  • Jacket length: 250 mm
  • Special size, non-DIN size
  • Typical application: product condenser for distillate separations
  • Pack unit: 1
TRUSTED_SUSTAINABILITY
Specifications
Material Borosilicate Glass (3.3)
Length (Metric) Jacket 250 mm
Type Allihn Condenser