Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences DURAN™ Aspirator Laboratory Bottle, GL 45 Thread Neck, GL 32 Thread Side Arm
SDP

Catalog No. 02900131 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
10,000 mL
1000 mL
2000 mL
5000 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
02-900-131 5000 mL
02-900-129 1000 mL
02-900-132 10,000 mL
02-900-130 2000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 02-900-131 Supplier DWK Life Sciences Supplier No. 247037309
Add to Cart
Add to Cart

Aspirator laboratory bottle with screw closure and PTFE key stopcock

Aspirator laboratory bottle with screw closure, complete with PTFE key stopcock and GL 45 red PBT cap.

  • Supplied ready to use
  • DURAN™ glass and PTFE key stopcock outlet
  • Made of virtually inert, chemically resistant DURAN™ glass
  • Fully compatible with all the GL 45 threads and therefore highly versatile
  • Tabulator with GL 32 thread
  • Easy-to-read scale with graduation marks to estimate the content
  • Fired-on, highly durable white ceramic
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 5000 mL
For Use With (Application) Storage and dispensing of liquids and reagents
Material Borosilicate Glass (3.3), Silicone, PTFE
No. per Pack 1/Pk.
Neck Finish Thread Size GL 45
Diameter (Metric) 182 mm
Autoclavable Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.