Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences DURAN™ GL 32 Screw Caps for Laboratory Glass Bottles
SDP

Catalog No. 09841263 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Blue
Red
Material:
PBT
Polypropylene
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Material
09-841-263 Blue Polypropylene
09-841-269 Red PBT
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-841-263 Supplier DWK Life Sciences Supplier No. 292391907
Add to Cart
Add to Cart

Distinguishing media types is simplified.

  • Available in blue, green, yellow and grey
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Material Polypropylene
Thread Size (Metric) GL 32
Product Type Screw Cap
For Use With (Equipment) Laboratory glass bottles
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.