Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences DURAN™ GL 45 Pouring Rings For Screw Caps
SDP

Catalog No. 09841241 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Blue
Gray
Green
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
09-841-241 Green
09-841-240 Blue
09-841-242 Gray
09-841-244 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 09-841-241 Supplier DWK Life Sciences Supplier No. 1089911
Add to Cart
Add to Cart

Distinguishing media types is simplified.

  • Available in blue, green, yellow and grey
TRUSTED_SUSTAINABILITY

Specifications

Color Green
Height (Metric) With Closure 4mm
Material Polypropylene
Product Type Pouring Ring
For Use With (Equipment) Screw Cap
Autoclavable Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.