Learn More
Description
- Available in blue, green, yellow and grey
Specifications
Specifications
| Color | Gray |
| Height (Metric) With Closure | 4mm |
| Material | Polypropylene |
| Product Type | Pouring Ring |
| For Use With (Equipment) | Screw Cap |
| Autoclavable | Autoclavable |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
