Learn More
Sigma Aldrich Fine Chemicals Biosciences Borosilicate Glass® Laboratory Bottle with Graduations | 2000 ML | Clear
Supplier:  Sigma Aldrich Fine Chemicals Biosciences DWK2180586041EA
Encompass
The borosilicate glass 3.3 DURAN PROTECT LABORATORY BTL provides chemical and thermal resistance, high mechanical strength, and thermal shock resistance. The wide mouth enables easy cleaning. Graduations up to 2000ml allow for precise volume measurements. This autoclavable laboratory bottle can withstand temperatures up to 500°C and meets ISO 9001 standards as well as USP and EP pharmacopeia requirements, making it suitable for the storage, mixing, heating, sterilization, and transport of laboratory chemicals and samples in a variety of laboratory settings including analytical, biological, chemical, clinical, environmental, food, industrial, medical, pharmaceutical, and scientific.
Catalog No. NC2339324
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.