Learn More
DWK Life Sciences DURAN™ YOUTILITY™ Bottle Tag, GL 45, Silicone Rubber, Assorted
Shop All DWK Life Sciences Products

Description
The colourful GL 45 Bottle Tags offer an elegant solution to the marking of laboratory bottles. The Bottle Tags can be easily and securely attached around the neck of the YOUTILITY™ bottle, even with the screw cap in place. The Bottle Tags feature a graspable tab that makes them easy to remove with bare or gloved hands.
- Available in different colours for marking of laboratory bottles and personalization
- The elastic Bottle Tags can be easily and securely attached around the neck, even with the screw cap in place
- Feature a graspable tab that makes them easy to remove with bare or gloved hands
- Chemically robust, heat resistant and totally reusable
- Fully compatible with all the GL 45 threads and therefore highly versatile
Specifications
Specifications
| Quantity | 16/Pk. |
| Color | Assorted |
| Material | Silicone |
| For Use With (Application) | color coding and identification |
| Style | GL 45 Bottle, Autoclavable |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.