Learn More
Medchemexpress LLC HY-P9919 1mg Medchemexpress, Durvalumab CAS:1428935-60-7 Purity:>98%
Supplier: Medchemexpress LLC HYP99191MG
Medchemexpress, HY-P9919 1mg Durvalumab CAS:1428935-60-7 Durvalumab (MEDI 4736) is an humanized anti- PD-L1 monoclonal antibody [1] . Durvalumab (MEDI4736) completely blocks the binding of PD-L1 to both PD-1 and CD80 , with IC 50 s of 0.1 and 0.04 nM, respectively [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.