Learn More
Invitrogen™ DUS2L Polyclonal Antibody

Description
Antibody detects endogenous levels of total DUS2L.
Specifications
Specifications
| Antigen | DUS2L |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | DUS2 |
| Gene Accession No. | Q9D7B1, Q9NX74 |
| Gene Alias | 2310016K04Rik; Dihydrouridine synthase 2; dihydrouridine synthase 2-like, SMM1 homolog; Dus2; Dus2l; hDUS2; RGD1305612; SMM1; SMM1 homolog; tRNA-dihydrouridine synthase 2-like; tRNA-dihydrouridine(20) synthase (NAD(P)(+)); tRNA-dihydrouridine(20) synthase [NAD(P)+]-like; Up-regulated in lung cancer protein 8; URLC8 |
| Gene Symbols | DUS2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.