Learn More
Invitrogen™ DUSP13 Polyclonal Antibody

Description
Suggested positive control: human skeletal muscle lysate. 93% sequence identity with Mouse.
Specifications
Specifications
| Antigen | DUSP13 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.2% gelatin and 0.05% sodium azide |
| Gene | DUSP13 |
| Gene Accession No. | Q6B8I1, Q9UII6 |
| Gene Alias | BEDP; Branching-enzyme interacting DSP; branching-enzyme interacting dual-specificity protein phosphatase; Dsp13-like protein; dual specificity phosphatase 13; Dual specificity phosphatase SKRP4; dual specificity protein phosphatase 13; Dual specificity protein phosphatase 13 isoform A; Dual specificity protein phosphatase 13 isoform B; dual specificity protein phosphatase isoform MDSP; dual specificity tyrosine phosphatase TS-DSP6; dual-specificity tyrosine phosphatase TS-DSP6; DUSP13; Dusp13a; DUSP13B; FLJ32450; Gm1203; LMW-DSP6; Mdsp; muscle-restricted DSP; muscle-restricted dual specificity phosphatase; muscle-restricted dual specificity phosphatase; testis and skeletal muscle-specific dual specificity phosphatase; RP11-487I5.6; SKRP4; testis- and skeletal muscle-specific DSP; testis and skeletal muscle-specific dual specificity phosphatase; Testis- and skeletal-muscle-specific DSP; TMDP; TS-DSP6 |
| Gene Symbols | DUSP13, DUSP13B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.