Learn More
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
Specifications
| Accession Number | AAH02671 |
| For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
| Formulation | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
| Gene ID (Entrez) | 1846 |
| Molecular Weight (g/mol) | 69.08 |
| Name | DUSP4 (Human) Recombinant Protein (P01) |
| pH Range | 8 |
| Preparation Method | In vitro wheat germ expression system |
| Purification Method | Glutathione Sepharose 4 Fast Flow |
| Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.