Learn More
Essendant DUSTER10OZTWIN-PACKBE
Supplier:  Essendant END11407
Encompass_Preferred
High-performance compressed air duster removes dust and lint from electronics Flexible extension tube provided for tight spaces and pinpoint cleaning Contains bitterant to discourage inhalant abuse Volume Capacity- 10 oz Product Packaging- Can Refillable (Yes/No)- No Propellant- 152A Liquefied Gas
Catalog No. 50-000-35502
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.