Learn More
Kemtec™ Dusting for Fingerprints Kit
SCI_ED
Catalog No. S27539 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Age:
12 to 18
Catalog No. Age
S27539 12 to 18
1 options
Catalog No. S27539 Supplier Kemtec™ Supplier No. 10512
Add to Cart
Add to Cart
Kemtec™ Dusting for Fingerprints Kit features advanced techniques using carbon, aluminum, magnetic, and fluorescent powders, magnetic applicators, fiberglass brushes, and camel fingerprinting brushes.
  • Teacher-developed three station design is efficient and cost effective.
  • Students perfect their techniques as they prepare fingerprint evidence.
  • Students use an inquiry approach to determine the best methodologies to stabilize, dust, lift, and develop prints from light, dark, smooth, textured, porous, and non-porous plastics, metals, wood, and vinyl surfaces.
  • Students use what they have learned to analyze and compare the minutia of print evidence to solve a crime at the Museum of the Modern Detective.
  • Allow 4 to 6 hours of lab time to complete all five experiments.
  • Includes supplies for 24 students working in teams, complete instruction manual, reproducible student procedures, and assessment materials.
  • Hood, UV light source, and goggles required.
  • Requires a special permit for international shipping.
  • Meets National Standards for Grade Levels 7 to 12.
Specifications
Grade 7 to 12
Age 12 to 18
Includes Instructor's Manual, Charcoal Print Powder, Aluminum Print Powder, Camel Hair Brushes, Fiberglass Fingerprint Brushes, Black Magnetic Print , Powder, Fluorescent Magnetic Print Powder, Magnetic Applicator Brushes, Magnetic Applicator Replacement Shields, Ink pads, Wood Flooring Samples, Vinyl Flooring Samples, Plastic Containers, Super Glue, Aluminum Foil, Plastic Spoons, Metal Spoons, White Notecards, Packing Tape, Magnifying Glasses
Weight (English) 10 lb.
Dimensions (L x W x H) 20 x 12 x 5 in.