Learn More
Bio-Serv™ Dustless Precision Pellets™ Purified Rodent Diet

Description
Precise size and weight pellets generally used in nutritional and behavioral studies
- Nutritionally complete purified formula
- Dustless and fracture free—superior performance in automatic feeders
- Every batch tested for hardness, weight and height
- Nutritionally assayed
Ingredients: Sucrose. Dextrose, Casein, Corn Oil, Mineral Mix, Cellulose, Corn Syrup, Calcium Silicate, Vitamin Mix, Magnesium Stearate, Choline Bitartrate, DL-Methionine, L-Cystine, Ascorbic Acid, Vitamin E Acetate, tBHQ, Chocolate Flavoring (F05301 and F0299 only)
Specifications
Specifications
| Product Type | Rodent Diet |
| Calories | 3.6 |
| Components | Sucrose. Dextrose, Casein, Corn Oil, Mineral Mix, Cellulose, Corn Syrup, Calcium Silicate, Vitamin Mix, Magnesium Stearate, Choline Bitartrate, DL-Methionine, L-Cystine, Ascorbic Acid, Vitamin E Acetate, tBHQ |
| Flavor | No Added Flavor |
| Format | Pellet |
| For Use With (Application) | Nutritional and behavioral studies |
| Particle Shape | Pellet |
| Quantity | 0.3 g |
| Species | Rodent |
| Storage Requirements | Cool dry place |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.