Learn More
Invitrogen™ DVL3 Polyclonal Antibody

Description
Antibody detects endogenous levels of total DVL3.
Specifications
Specifications
| Antigen | DVL3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide |
| Gene | DVL3 |
| Gene Accession No. | Q61062, Q92997 |
| Gene Alias | dishevelled 3 (homologous to Drosophila dsh); dishevelled 3, dsh homolog; dishevelled 3, dsh homolog (Drosophila); dishevelled segment polarity protein 3; dishevelled, dsh homolog 3; dishevelled-3; DRS3; DSH homolog 3; DVL3; dvl-3; I79_017820; KIAA0208; OTTHUMP00000210778; Segment polarity protein dishevelled homolog DVL-3; Segment polarity protein dishevelled-like DVL-3 |
| Gene Symbols | DVL3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.