Learn More
Selleck Chemical LLC DX3-213B-E1139-5MG
SDP
Supplier:  Selleck Chemical LLC E11395MG
Encompass_Preferred
DX3-213B is a highly potent orally active oxidative phosphorylation (OXPHOS) complex I inhibitor with IC50 of 3 6 nM and impairs ATP generation with IC50 of 11 nM
Catalog No. 50-313-4653