Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ DXD Antibody (1A1), R&D Systems™
SDP

Catalog No. NB777655
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB777655 100 μg
NB777283 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB777655 Supplier R&D Systems™ Supplier No. NBP348398100UG

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Rabbit Monoclonal Antibody

DXD Monoclonal antibody specifically detects DXD in Non-species specific samples. It is validated for ELISA

Specifications

Antigen DXD
Applications ELISA
Classification Monoclonal
Clone 1A1
Conjugate Unconjugated
Dilution ELISA 1:5000-10000
Formulation PBS, pH 7.4, 5-8% Trehalose
Gene Alias 1599440-33-1, Exatecan derivative for ADC
Host Species Rabbit
Immunogen The exact amino acid sequence is proprietary.
Purification Method Antigen Affinity-purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Reconstitution Dissolve the lyophilized antibody in sterile and low endotoxin water. The recommend volume of water is lot specific. Please refer to the instructions printed on the product vial or contact technical support.
Target Species Non-species specific
Content And Storage Store at -20°C in powder form. Store at -80°C once reconstituted.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.