Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ DY131
GREENER_CHOICE

Catalog No. 226610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-661-0 10 mg
22-665-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-661-0 Supplier Tocris Bioscience™ Supplier No. 2266/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Agonist selective for estrogen-related receptors ERRβ and ERRγ

Novel selective agonist at estrogen-related receptors ERRβ and ERRγ. Displays minimal activity at ERRα, ERα and ERβ at concentrations up to 30 μM.

Chemical Identifiers

CAS 95167-41-2
Molecular Formula C18H21N3O2

Specifications

Quantity 10 mg
Formula Weight 311.38
Purity >99%
Molecular Weight (g/mol) 311.385
Chemical Name or Material DY131
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.