Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dynalon™ Tote Box Cover
SCI_ED

Catalog No. S04527 Shop All Dynalab Corporation Products
Change view
Click to view available options
Color:
Natural
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S04527 Natural
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is intended for educational purposes only.
Catalog No. S04527 Supplier Dynalon™ Supplier No. 4105350001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is intended for educational purposes only.

For sterilizing tote box.

  • Made of natural polypropylene
  • Chemically resistant so will not rust or corrode
  • Is autoclavable
  • Dimensions (L x W x H): 12.25 x 7.75 x 0.5 in. (31.1 x 19.7 x 1.3cm)

Specifications

Material Polypropylene
Color Natural
Height (English) 0.5 in.
Height (Metric) 1.3 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.