Learn More
Dyclonine hydrochloride
Catalog No. AAJ6133006
Change view
Click to view available options
Quantity:
5 g
Catalog No. Quantity
AAJ6133006 5 g
1 options
Catalog No. AAJ6133006 Supplier Thermo Scientific Chemicals Supplier No. J6133006
Dyclonine hydrochloride is used for the temporary relief of the following occasional mouth and throat symptoms: pain, minor irritation, sore mouth and sore throat. It is a sodium channel blocker and a HSP90 chaperone protein inhibitor. It shows local anesthetic effects in vivo and is orally active.
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 5 g