Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dynalon™ Dynalon Hydrometer Racks
SCI_ED

Catalog No. S37578 Shop All Dynalab Corporation Products
Change view
Click to view available options
Size:
Short
Tall
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
S37578 Tall
S37579 Short
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S37578 Supplier Dynalon™ Supplier No. 1032150000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed to safeguard glass and plastic hydrometers during storage and transport.

  • Made from polypropylene
  • Three tiered rack with handles and stable base
  • Top two plates have 1 in. diameter holes; bottom plate has wells which taper from 1 to 0.37 in.
  • Available in tall and short sizes

Specifications

Material Polypropylene
Positions 18
For Use With (Application) To safeguard glass and plastic hydrometers during storage and transport.
Size Tall
Dimensions (L x W x H) 21 x 21.5 x 25 cm (8.25 x 8.5 x 9.87 in.)
Height (English) Shelf 7 in.
Height (Metric) Shelf 17.78 cm
Includes Handles and stable base
Product Type Hydrometer Rack
Shelves 3 Tiers
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.