Learn More
Dynamic Diagnostics Safety Shields
Magnetic or clamp-on splash shields with gooseneck arm for exact angle positioning
$204.00 - $209.00
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
22-899-024
|
Dynamic Diagnostics
300017 |
Magnetic Safety Shield |
Each for $204.00
|
|
|||||
22-899-025
|
Dynamic Diagnostics
300018 |
Clamp-on Safety Shield |
Each for $209.00
|
|
|||||
Description
- Exact angle positioning
- 12 x 8.5 in.
- Clear polycarbonate
- UV protection from 200-360 nanometers
- Available as magnetic base or a rubber-coated, clamp-on base
Specifications
1 Ea. |