Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Dynamics Carts
SDPSCI_ED

Catalog No. S041375 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
6 to 15
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S041375 6 to 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

Prop 65 - Cancer and Reproductive Harm

Catalog No. S041375 Supplier United Scientific™ Supplier No. DNCART2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Prop 65 - Cancer and Reproductive Harm

Ideal for demonstrating experiments in conservation, velocity, collisions, and momentum.

  • Ideal for demonstrating experiments in: Conservation, Velocity, Elastic collisions, Acceleration, Momentum
  • Included are two impact carts and two spring steel bumpers with rubber stopper assemblies
  • Includes activity guide
  • Each cart features a well for additional masses (not included)

Specifications

Product Type Collision Cart
Grade 6 to 15
For Use With (Application) Demonstrating Experiments in Conservation, Velocity, Elastic Collisions, Acceleration and Momentum.
Includes Two impact carts and two spring steel bumpers with rubber stopper assemblies
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.