Learn More
Invitrogen™ Dynamin 2 Polyclonal Antibody

Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
Specifications
Specifications
| Antigen | Dynamin 2 |
| Applications | Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.9 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene Alias | b2b2159Clo; CMT2M; CMTDI1; CMTDIB; DI-CMTB; Dnm2; DYIIAAB; Dyn2; dynamin 2; dynamin II; Dynamin UDNM; Dynamin-2; DYNII; LCCS5; Udnm |
| Host Species | Rabbit |
| Immunogen | The immunogen used to generate this antibody corresponds to zebrafish Dnm2. |
| Purification Method | Antigen Affinity Chromatography |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.