Learn More
Dynamitin (1-401 AA) Mouse anti-Human, Mouse, Clone: 2B9A5, Proteintech
Shop All Proteintech Group Inc Products

Description
This antibody is against the N terminal 86aa of full length p50 dynamitin.
Specifications
Specifications
| Antigen | Dynamitin (1-401 AA) |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 2B9A5 |
| Concentration | 1.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | DCTN2 |
| Gene Accession No. | Q13561, Q99KJ8 |
| Gene Alias | DCTN2, dynactin 2, dynactin 2 (p50), Dynactin subunit 2, Dynamitin |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.